Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NeuroD 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5688
Description
Anti-NeuroD 2 Antibody detects level of NeuroD 2 and has been published and validated for use in immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin) and western blotting.
Specifications
NeuroD 2 | |
Polyclonal | |
Affinity purified immunoglobulin. Precipitated antibody in a solution of 50% saturated ammonium sulfate and PBS containing no preservatives. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat |
Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
NEUROD2; NDRF; NeuroD2; MGC26304 | |
Synthetic peptide, amino acids 23-39 of human NeuroD2. | |
100 μg | |
NM_006160.3 | |
-70°C for up to 6 months, Avoid Freeze/Thaw Cycles |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction