Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

neurogenic differentiation 1, Mouse, Clone: 3H8, Abnova™

Catalog No. 89019066 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-019-066 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-019-066 Supplier Abnova Corporation Supplier No. H00004760M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant NEUROD1.

This gene encodes a member of the NeuroD family of basic helix-loop-helix (bHLH) transcription factors. The protein forms heterodimers with other bHLH proteins and activates transcription of genes that contain a specific DNA sequence known as the E-box. It regulates expression of the insulin gene, and mutations in this gene result in type II diabetes mellitus. [provided by RefSeq

Sequence: QDMPPHLPTASASFPVHPYSYQSPGLPSPPYGTMDSSHVFHVKPPPHAYSAALEPFFESPLTDCTSPSFDGPLSPPLSINGNFSFKHEPSAEFEKNYAFT

Specifications

Antigen neurogenic differentiation 1
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 3H8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant NEUROD1.
Formulation PBS with no preservative; pH 7.4
Gene NEUROD1
Gene Accession No. BC009046
Gene Alias BETA2/BHF-1/NEUROD/bHLHa3
Gene Symbols NEUROD1
Host Species Mouse
Immunogen NEUROD1 (AAH09046, 201 a.a. ∼ 300 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neurobiology
Primary or Secondary Primary
Gene ID (Entrez) 4760
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.