Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEUROD2, Rabbit, MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002907 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-907 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-907 Supplier Abnova Corporation Supplier No. H00004761D01
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human NEUROD2 protein.

This gene encodes a member of the neuroD family of neurogenic basic helix-loop-helix (bHLH) proteins. Expression of this gene can induce transcription from neuron-specific promoters, such as the GAP-43 promoter, which contain a specific DNA sequence known as an E-box. The product of the human gene can induce neurogenic differentiation in non-neuronal cells in Xenopus embryos, and is thought to play a role in the determination and maintenance of neuronal cell fates. [provided by RefSeq

Sequence: MLTRLFSEPGLLSDVPKFASWGDGEDDEPRSDKGDAPPPPPPAPGPGAPGPARAAKPVPLRGEEGTEATLAEVKEEGELGGEEEEEEEEEEGLDEAEGERPKKGGPKKRKMTKARLERSKLRRQKANARERNRMHDLNAALDNLRKVVPCYSKTQKLSKIETLRLAKNYIWALSEILRSGKRPDLVSYVQTLCKGLSQPTTNLVAGCLQLNSRNFLTEQGADGAGRFHGSGGPFAMHPYPYPCSRLAGAQCQAAGGLGGGAAHALRTHGYCAAYETLYAAAGGGGASPDYNSSEYEGPLSPPLCLNGNFSLKQDSSPDHEKSYHYSMHYSALPGSRPTGHGLVFGSSAVRGGVHSENLLSYDMHLHHDRGPMYEELNAFFHN

Specifications

Antigen NEUROD2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human NEUROD2 protein.
Formulation No additive
Gene NEUROD2
Gene Accession No. BC022481.1
Gene Alias MGC26304/NDRF/bHLHa1
Gene Symbols NEUROD2
Host Species Rabbit
Immunogen NEUROD2 (AAH22481.1, 1 a.a. ∼ 382 a.a) full-length human protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4761
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.