Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

neurogenic differentiation 6, Mouse, Clone: 3B3, Abnova™

Catalog No. 89001338 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-338 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-338 Supplier Abnova Corporation Supplier No. H00063974M06
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant NEUROD6.

NEUROD6 is a member of the NEUROD (NEUROD1; MIM 601724) family of basic helix-loop-helix (bHLH) transcription factors (Guo et al., 2002 [PubMed 12357074]).[supplied by OMIM

Sequence: GQGGEAAHHTRSPYSTFYPPYHSPELTTPPGHGTLDNSKSMKPYNYCSAYESFYESTSPECASPQFEGPLSPPPINYNGIFSLKQEETLDYGKNYNYGMH

Specifications

Antigen neurogenic differentiation 6
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3B3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant NEUROD6.
Formulation PBS with no preservative; pH 7.4
Gene NEUROD6
Gene Accession No. NM_022728
Gene Alias Atoh2/MATH2/Math-2/NEX1M/bHLHa2
Gene Symbols NEUROD6
Host Species Mouse
Immunogen NEUROD6 (NP_073565.2, 191 a.a. ∼ 290 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 63974
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.