Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofilament 200 kDa Mouse anti-Human, Mouse, Rat, Clone: RT97, MilliporeSigma™

Catalog No. MAB5262MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5262MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5262MI Supplier MilliporeSigma Supplier No. MAB5262
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Neurofilament 200 kDa Antibody, clone RT97 is an antibody against Neurofilament 200 kDa for use in immunohistochemistry, immunohistochemistry (paraffin) and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurofilament 200 kDa
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone RT97
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in 0.02MPhosphate buffer, 0.25MNaCl with 0.1% sodium azide.
Gene Accession No. NM_021076.2
Gene Symbols NEFH, NFH, NF-H, KIAA0845
Host Species Mouse
Immunogen Rat brain protein preparation
Purification Method Protein A purfied
Quantity 50 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.