Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofilament 70 kDa Mouse anti-Bovine, Human, Mouse, Pig, Rat, Clone: DA2, MilliporeSigma™

Catalog No. MAB1615MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
300 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1615MI 300 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1615MI Supplier MilliporeSigma Supplier No. MAB1615
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Neurofilament 70 kDa Antibody, clone DA2 is validated for use in western blot, immunocytochemistry, immunohistochemistry for the detection of Neurofilament 70 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurofilament 70 kDa
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone DA2
Conjugate Unconjugated
Formulation Liquid. Contains no preservative.
Gene Accession No. NM_006158.2
Gene Symbols NEFL, CMT1F, NF-L, NF68, CMT2E, NFL
Host Species Mouse
Immunogen Enzymatically dephosphorylated pig neurofilaments.
Quantity 300 μL
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Pig, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.