Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurofilament H (200 kDa) Rabbit anti-Bovine, Human, Mouse, Pig, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB1989
Description
Detect Neurofilament H (200 kDa) using this Anti-Neurofilament H (200 kDa) Antibody, C-terminus validated for use in immunohistochemistry and western blotting.
Specifications
Neurofilament H (200 kDa) | |
Polyclonal | |
Neat rabbit antisera, no preservatives. | |
Rabbit | |
Unpurified | |
Primary | |
Bovine, Human, Mouse, Pig, Rat | |
Serum |
Immunohistochemistry, Western Blot | |
Unconjugated | |
NEFH; NFH; NF-H; KIAA0845 | |
Recombinant fusion protein containing the extreme C-terminus of rat NF-H, prepared from E. coli inclusion bodies by dissolution in urea followed by DEAE-cellulose ion exchange chromatography. Immunogen does not include the lysine-serine-proline | |
50 μL | |
NM_021076.2 | |
−20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction