Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurofilament H (200 kDa) Rabbit anti-Human, Mouse, Bovine, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABN76
Description
This Anti-Neurofilament H (200 kDa) Antibody is validated for use in western blot, immunohistochemistry (paraffin) for the detection of Neurofilament H (200 kDa).
Specifications
Neurofilament H (200 kDa) | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Bovine, Human, Mouse, Rat |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
NEFH | |
GST-tagged recombinant protein corresponding to bovine Neurofilament H. | |
100 μg | |
NP_066554.2 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction