Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofilament H (200 kDa) Rabbit anti-Avian, Mammals, Reptile, Polyclonal, MilliporeSigma™

Catalog No. AB1991MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1991MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1991MI Supplier MilliporeSigma Supplier No. AB1991
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Neurofilament H (200 kDa) using this Anti-Neurofilament H (200 kDa) Antibody, lysine-serine-proline repeat validated for use in immunohistochemistry (paraffin), western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurofilament H (200 kDa)
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Neat rabbit antisera.
Gene Symbols NEFH; NFH; NF-H; KIAA0845
Host Species Rabbit
Immunogen E. Coli recombinant fusion protein containing 37 lysine-serine-proline repeats of rat NF-H.
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_021076.2
Target Species Avian, Mammalia, Reptile
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.