Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofilament H Mouse anti-Rat, Human, Pig, AlexaFluor™488, Clone: NE14, MilliporeSigma™

Catalog No. MAB5256XMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
40 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5256XMI 40 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5256XMI Supplier MilliporeSigma Supplier No. MAB5256X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Neurofilament H Antibody, clone NE14, Alexa Fluor 488 Conjugated is an antibody against Neurofilament H for use in immunohistochemistry and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurofilament H
Applications Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone NE14
Conjugate Alexa Fluor 488
Formulation Purified immunoglobulin in 0.02M Phosphate buffer, 0.25M NaCl containing 0.1% sodium azide.
Gene Accession No. NP_066554.2
Gene Symbols NEFH, NF-H, NFH
Host Species Mouse
Immunogen Purified neurofilament polypeptides (Debus et al., 1983).
Purification Method Protein G purfied
Quantity 40 mg
Primary or Secondary Primary
Target Species Human, Pig, Rat
Content And Storage 2-8°C for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.