Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofilament H Chicken anti-Bovine, Human, Mouse, Pig, Rat, Feline, Polyclonal, MilliporeSigma™

Catalog No. AB5539MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5539MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5539MI Supplier MilliporeSigma Supplier No. AB5539
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Anti-Neurofilament H Antibody is an antibody against Neurofilament for use in immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin) and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurofilament H
Applications Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified chicken IgY in PBS containing 5mM sodium azide.
Gene Symbols NEFH; NF-H; NFH; KIAA0845
Host Species Chicken
Immunogen Purified bovine NF-H.
Purification Method Ammonium sulfate precipitation
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_021076.2
Target Species Bovine, Feline, Human, Mouse, Pig, Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.