Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofilament L Rabbit anti-Human, Mouse, Rat, Feline, Canine, Pig, Bovine, Polyclonal, MilliporeSigma™

Catalog No. AB9568MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9568MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9568MI Supplier MilliporeSigma Supplier No. AB9568
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Neurofilament L using this Anti-Neurofilament L Antibody validated for use in immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin) and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurofilament L
Applications Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit serum. Contains no preservative.
Gene Symbols NEFL; CMT1F; NF-L; NF68; CMT2E; NFL
Host Species Rabbit
Immunogen Purified porcine NF-L
Purification Method Unpurified
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_006158.2
Target Species Bovine, Canine, Feline, Human, Mouse, Pig, Rat
Content And Storage −20°C for up to 12 months
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.