Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofilament M (145 kDa) Rabbit anti-Bovine, Chicken, Human, Mouse, Pig, Rat, Quail, Rabbit, Reptile, Polyclonal, MilliporeSigma™

Catalog No. AB1987MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1987MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1987MI Supplier MilliporeSigma Supplier No. AB1987
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Neurofilament M (145 kDa) Antibody, C-terminus detects level of Neurofilament M (145 kDa) and has been published and validated for use in immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin) and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurofilament M (145 kDa)
Applications Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Neat rabbit antiserum containing no preservative.
Gene Symbols NEFM; NF-M; NEF3; NFM
Host Species Rabbit
Immunogen Recombinant fusion protein containing the C-terminal 168 amino acids of rat NF-M
Purification Method Unpurified
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_001105541.1
Target Species Bovine, Chicken, Human, Mouse, Pig, Quail, Rabbit, Rat, Reptile
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.