Learn More
Neurofilament M Chicken anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Neurofilament M |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Chicken IgY. Liquid containing ∼5mM sodium azide. |
| Gene Symbols | NEFM; NF-M; NEF3; NFM |
| Host Species | Chicken |
| Immunogen | Recombinant C-terminal extension of rat NF-M, the so-called KE segment. |
| Quantity | 50 μL |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.