Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofilament NF-H Rat anti-Human, Hamster, Monkey, Rat, Clone: TA51, MilliporeSigma™

Catalog No. MAB5448MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5448MI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5448MI Supplier MilliporeSigma Supplier No. MAB5448
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Neurofilament NF-H Antibody, clone TA51 is an antibody against Neurofilament NF-H for use in ELISA, immunoprecipitation, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurofilament NF-H
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone TA51
Conjugate Unconjugated
Formulation Unpurified tissue culture supernatant from a perfusion system, filtered through a 0.2μ membrane prior to vialing. Product contains 20%FBS and Ciprofloxacin at final concentration of 10μg/mL.
Gene Accession No. NM_021076.2
Gene Symbols NEFH, NFH, NF-H, KIAA0845
Host Species Rat
Quantity 1 mL
Primary or Secondary Primary
Target Species Hamster, Human, Monkey, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.