Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurogenin-3 (rabbit) Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB10535MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB10535MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB10535MI Supplier MilliporeSigma Supplier No. AB10535
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Neurogenin-3 (rabbit) using this Anti-Neurogenin-3 (rabbit) Antibody validated for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurogenin-3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Unpurified rabbit polyclonal serum containing 0.05% sodium azide.
Gene Symbols NEUROG3; ATOH5; Atoh5; ngn3; NGN3; MATH4B; Math4B
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to the N-terminus of mouse Neurogenin-3.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_033849
Target Species Mouse
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.