Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neurogranin Rabbit anti-Bovine, Chicken, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5620
Description
This Anti-Neurogranin Antibody is validated for use in ELISA, immunocytochemistry, immunohistochemistry, immunoprecipitation, western blotting for the detection of Neurogranin.
Specifications
Neurogranin | |
Polyclonal | |
Purified immunoglobulin. Purified by ammonium sulfate precipitation. Liquid in PBS with 0.02% sodium azide as a preservative. | |
Rabbit | |
Ammonium sulfate precipitation | |
Primary | |
Bovine, Chicken, Mouse, Rat | |
Purified |
ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
NRGN; neurogranin; Ng; Neurogranin; RC3; hng | |
Recombinant rat Neurogranin (complete sequence). | |
100 μL | |
NM_006176.1 | |
−20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction