Learn More
Neuron Specific Enolase (NSE) Chicken anti-Human, Rat, Mouse, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Neuron Specific Enolase |
| Applications | Immunocytochemistry, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified chicken IgY. Two different antibodies were combined to make this product. Liquid in PBS, pH 7.2 with 0.02% sodium azide. |
| Gene Symbols | ENO2; NSE; Gamma-enolase; EC 4.2.1.11 |
| Host Species | Chicken |
| Immunogen | Synthetic peptides from human/rat/mouse Neuron Specific Enolase. |
| Purification Method | Affinity Purified |
| Quantity | 500 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.