Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuron Specific Enolase Mouse anti-Human, Clone: 5E2, MilliporeSigma™

Catalog No. MAB324MI Shop All MilliporeSigma Products
missing translation for 'changeView'
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB324MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. MAB324MI Supplier MilliporeSigma missing translation for 'supplierNo' MAB324
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Neuron Specific Enolase Antibody, clone 5E2 detects level of Neuron Specific Enolase and has been published and validated for use in ELISA, western blot, immunocytochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuron Specific Enolase
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 5E2
Conjugate Unconjugated
Formulation Liquid in PBS pH 7.4 containing 0.1% sodium azide.
Gene Accession No. NM_001975.2
Gene Symbols ENO2, NSE, Gamma-enolase, EC 4.2.1.11
Host Species Mouse
Immunogen Neuron-specific enolase from human brain.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for up to 12 months under sterile conditions.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.