Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuron-Specific Nuclear Protein Mouse anti-Avian, Chicken, Ferret, Human, Mouse, Pig, Rat, Primate, Salamander, Vertebrates, Clone: A60, MilliporeSigma™

Catalog No. IHCR10016MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
6 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IHCR10016MI 6 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IHCR10016MI Supplier MilliporeSigma Supplier No. IHCR10016
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Neuron-Specific Nuclear Protein Antibody, prediluted, clone A60 detects level of Neuron-Specific Nuclear Protein and has been published and validated for use in immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuron-Specific Nuclear Protein
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone A60
Conjugate Unconjugated
Formulation Liquid diluted in PBS, pH 7.2 with stabilizers, 0.2% Tween 20, and 0.1% ProClin 300 as preservative.
Host Species Mouse
Immunogen Purified cell nuclei from mouse brain.
Quantity 6 mL
Primary or Secondary Primary
Target Species Avian, Chicken, Ferret, Human, Mouse, Pig, Primate, Rat, Salamander, Vertebrates
Content And Storage 2-8°C.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.