Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuronal pentraxin 2 Rabbit anti-Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB15520
Description
Detect Neuronal pentraxin 2 using this Anti-Neuronal pentraxin 2 Antibody validated for use in ELISA and western blotting.
Specifications
Neuronal pentraxin 2 | |
Polyclonal | |
Liquid in 100mM Tris, pH 7.5, with 0.2% BSA and 0.05% sodium azide. | |
Rabbit | |
Immunoaffinity purified | |
Primary | |
Mouse | |
Purified |
ELISA, Western Blot | |
Unconjugated | |
NPTX2; NP-II; NP2; NARP | |
Synthetic peptide near the N-terminus of mouse NPX2 | |
50 μg | |
NM_002523.1 | |
−20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction