Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuronal Pentraxin Mouse, Unlabeled, Clone: 22, BD

Catalog No. BDB610369 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610369 50 μg
BDB610370 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610369 Supplier BD Biosciences Supplier No. 610369
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neuronal pentraxin (NP) is a potential receptor for the snake venom toxin, taipoxin. The highest levels of NP mRNA are found in the cerebellum and the CA3 region of the hippocampus. More specifically, NP mRNA is expressed at high levels in Purkinje and granule neurons of the cerebellum and CA3 neurons of the hippocampus. Moderate levels are present in neurons of the hilus, dentate gyrus, CA1, cerebral cortex, medial habenula, anterior hypothalamic nuclei, olfactory cortex, and inferior olivary nuclei. NP mRNA is not expressed in glia of white or gray matter. It is thought that NP serves as a receptor for taipoxin and, after binding, NP-taipoxin is taken into presynaptic terminals. Alternatively, the binding of taipoxin by NP may represent a nonspecific mechanism to sequester toxins from synapses.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuronal Pentraxin
Applications Western Blot
Classification Monoclonal
Clone 22
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.