Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuropeptide Y2 Receptor Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB9358MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9358MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9358MI Supplier MilliporeSigma Supplier No. AB9358
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Neuropeptide Y2 Receptor Antibody is validated for use in immunohistochemistry for the detection of Neuropeptide Y2 Receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuropeptide Y2 Receptor
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in PBS with 0.05% sodium azide.
Gene Symbols NPY2R; NPY2-R
Host Species Rabbit
Immunogen Synthetic peptide from Neuropeptide Y2 Receptor.
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000910.2
Target Species Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.