Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Neurophysin 2/NP-AVP Clone: PS 41,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABN845
Description
Specifically detects Neurophysin 2/NP-AVP Clone: PS 41 in Rat samples, and it is validated for Electron Microscopy, ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmunoassay, Western Blotting
The neurophysin 1 and 2 (NPI and NPII) are carriers of peptide hormone oxytocin (OT) and vasopressin (AVP), respectively. Each NP and its associated hormone peptide are synthesized as one precursor protein, OT-NPI (UniProt P01179) encoded by the Oxt (or Ot) gene or AVP-NPII (UniProt P01186) encoded by the Avp gene in rat species. Post-translational cleavages remove the signal peptide and process the precursor into mature OT and NPI (aa 20-28 and aa 32-135, respectively, of OT-NPI) or AVP and NPII (aa 24-32 and aa 36-128, respectively, of AVP-NPII). OT is a potent hormone and neurotransmitter secreted within the pituitary gland. OT exerts its biological activity through its receptor OXTR (also known as OTR). OT is used clinically for labor induction and may exhibit therapeutic potential for treating social anxiety disorders. AVP functions primarily as an anti-diuretic and regulates the concentration of water, glucose, and salts in blood. AVP is secreted from the posterior pituitary, but exerts its biological activity in the kidney via the G protein-coupled receptor AVPR2 (V2R).
Specifications
Neurophysin 2/NP-AVP | |
Monoclonal | |
Unconjugated | |
P01186 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Clone PS 41 reacts with Vasopressin-neurophysin 2-copeptin precursor, neurophysin 2, but not Arg-vasopressin, copeptin, oxytocin, or neurophysin 1. | |
2°C to 8°C for one year from date of shipment | |
IgG2b κ |
Immunoassay, ELISA, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot | |
PS 41 | |
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Avp | |
KLH-conjugated acid soluble extract of rat (Sprague-Dawley) intermediate lobes. | |
100 μg | |
Neuroscience | |
NP_058688 | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction