Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuropilin-1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB9600MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9600MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9600MI Supplier MilliporeSigma Supplier No. AB9600
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Neuropilin-1 Antibody is an antibody against Neuropilin-1 for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuropilin-1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin in PBS containing 50% glycerol, 1mg/mL BSA and 0.05% sodium azide.
Gene Symbols NRP1; neuropilin-1; DKFZp781F1414; VEGF165R; NRP; DKFZp686A03134; CD304
Host Species Rabbit
Immunogen Synthetic peptide from the N-terminal CUB domain of human neuropilin-1.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_001024628.1; NM_001024629.1; NM_003873.4
Target Species Human
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.