Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuropilin-1 Rabbit anti-Rat, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB10521MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB10521MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB10521MI Supplier MilliporeSigma Supplier No. AB10521
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Neuropilin-1 Antibody is validated for use in western blot, immunohistochemistry, immunoprecipitation, immunofluorescence for the detection of Neuropilin-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuropilin-1
Applications Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit serum in buffer containing 0.05% sodium azide.
Gene Symbols NRP1; NRP; NP1; BDCA4; CD304; neuropilin-1
Host Species Rabbit
Immunogen This antibody is generated from rabbits immunized with a His-tagged partial recombinant rat Neuropilin-1.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_659566.1
Target Species Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.