Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurotensin Receptor 3 (NTR3) Mouse, Unlabeled, Clone: 48, BD

Catalog No. BDB612100 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612100 50 μg
BDB612101 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612100 Supplier BD Biosciences Supplier No. 612100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neurotensin (NT) is a neuropeptide that modulates dopaminergic transmission, triggers analgesic responses, induces hypotension, decreases gastric acid secretion, and activates lipid digestion. NT is a ligand for NT receptors, which include both G-protein coupled and non-G-protein coupled receptors. Neurotensin Receptor 3 (NTR3) is a non-G-protein coupled receptor that has a lumenal domain homologous to sorting proteins, and a short cytoplasmic tail homologous to mannose-6-phosphate/IGF-II receptor. NTR3 was also identified as sortilin/gp95, a component of GLUT4 vesicles in adipocytes that has been implicated in lipoprotein lipase degradation. NTR3 mRNA is expressed in brain, skeletal muscle, heart, and adipocytes. NTR3 mature protein has 44 N-terminal amino acid residues cleaved off, which may facilitate ligand binding to the receptor. Cellular localization of NTR3 is in the Golgi compartment and vesicles, as well as on the cell surface. In addition to roles in neuropeptide and lipoprotein degradation, NTR3 may also be involved in receptor protein sorting. Thus, NTR3 is a multifunctional transmembrane protein that acts as both a intracellular sorting receptor and extracellular ligand-binding receptor.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurotensin Receptor 3 (NTR3)
Applications Western Blot
Classification Monoclonal
Clone 48
Concentration 250μg/mL
Conjugate Unconjugated
Description NTR3
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Neurotensin Receptor 3 aa. 300-422
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.