Learn More
Neurotrophin 3 Rabbit anti-Chicken, Human, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Neurotrophin 3 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Protein A purified immunoglobulin. Lyophilized from PBS, no preservatives. Reconstitute with 500μL of sterile distilled water (1mg/mL). |
| Gene Symbols | NTF3; NT3; NGF2; HDNF; MGC129711; NGF-2; NT-3 |
| Host Species | Rabbit |
| Immunogen | An 11 amino acid peptide corresponding to the amino-terminal of mouse Neurotrophin 3 (NT3) coupled to BSA. The sequence is H-YAEHKSHRGEY-NH2. |
| Quantity | 500 μg |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.