Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurotrophin 4/5 Rabbit anti-Human, Mouse, Monkey, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1781SPMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1781SPMI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1781SPMI Supplier MilliporeSigma Supplier No. AB1781SP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Neurotrophin 4/5 Antibody is an antibody against Neurotrophin 4/5 for use in ELISA, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurotrophin 4/5
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Lyophilized from PBS, no preservatives. Reconstitute with 500μL of sterile distilled water (1mg/mL).
Gene Symbols NTF5; NT5; NT-4; NTF4; Neurotrophin-4; NT-5; NT-4/5; NT4
Host Species Rabbit
Immunogen Recombinant human Neurotrophin 4 (NT4).
Quantity 500 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_006179.3
Target Species Human, Monkey, Mouse, Rat
Content And Storage Lyophilized: −20-−70°C for up to 6 months. Reconstituted: −20-−70°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.