Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurturin Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5404MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5404MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5404MI Supplier MilliporeSigma Supplier No. AB5404
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Neurturin Antibody detects level of Neurturin and has been published and validated for use in immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurturin
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit serum. Lyophilized. Contains no preservative. Reconstitute with 50μL of sterile distilled water. Centrifuge if necessary to remove any residue.
Gene Symbols NRTN; NTN; neurturin
Host Species Rabbit
Immunogen Recombinant human Neurturin.
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_004558.2
Target Species Human, Mouse, Rat
Content And Storage Lyophilized: −20-−70°C for up to 6 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.