Learn More
Neutral amino acid transporter (ASCT2) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABN73
Description
Anti-Neutral amino acid transporter (ASCT2) Antibody detects level of Neutral amino acid transporter (ASCT2) and has been published and validated for use in western blot, immunohistochemistry (paraffin).
Specifications
Neutral amino acid transporter (ASCT2) | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
SLC1A5; M7V1; AAAT; R16; ATBO; M7VS1; RDR; ATB; ASCT2; RDRC | |
KLH-conjugated linear peptide corresponding to the N-terminus of Neutral amino acid transporter. | |
100 μg | |
NP_005619 | |
2-8°C for 12 months |