Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NF-ATc1 Mouse, Unlabeled, Clone: 7A6, BD

Catalog No. BDB556602 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556602 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556602 Supplier BD Biosciences Supplier No. 556602
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The NF-AT family of transcription factors are regulators of early immune response genes in T cells following their activation by CD40L, FasL and nterleukins including IL-2, -3, -4 and -5. NF-AT proteins function in both signal transduction and transcription control. The DNA binding, active NF-AT complex contains a cytoplasmic, Ca2+/calcineurin dependent, cyclosporin sensitive subunit, designated NF-ATc, which is present in the cytoplasm of resting cells. Upon cell activation, NF-ATc is dephosphorylated and thus activated by calcineurin, resulting in rapid translocation of NF-ATc to the nucleus. The active NF-AT complex also contains a nuclear component, NF-ATn. In vitro, NF-AT cooperates with the mitogenic transcription factor AP-1 to induce multiple cytokine genes. NF-ATc is encoded by four genes: NF-ATc1 (originally named NF-ATc), NF-ATc2 (originally named NF-ATp), NF-ATc3 (NF-AT3) and NF- ATc4 (NF-AT4). These proteins are differentially expressed in tissues, suggesting that each may activate distinct sets of genes. NF-ATc1 is expressed in lymphoid organs and is upregulated in activated T cell and thymocytes. NF-ATc1 is required for the development of T cells and has been shown to contribute to the production of IL-2 and the development of Th2 responses. 7A6 recognizes human and mouse NF-ATc1. The antibody recognizes a region between amino acids 197 to 304 of human NF-ATc1, which lies outside the region of homology between NF-ATc1 and NF-ATc2.3 Please note that in the original publication for the antibody (Northrop et al. 1994), the clone name 7A6 is not specifically identified.

Host Species: Mouse
Clone: 7A6
Isotype: IgG1
Cross Reactivity: Human, Mouse

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen NF-ATc1
Applications Western Blot
Classification Monoclonal
Clone 7A6
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.