Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFκB Mouse anti-Human, Mouse, Rat, Rabbit, Clone: 12H11, MilliporeSigma™

Catalog No. MAB3026MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3026MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3026MI Supplier MilliporeSigma Supplier No. MAB3026
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-NFκB Antibody, p65 subunit, active subunit, clone 12H11 (Mouse Monoclonal Antibody) validated in EMSA, flow cytometry, immunocytochemistry, immunofluoresence, immunohistochemistry, immunohistochemistry (paraffin), western blotting to detect NFκB also known as Rel A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFκB
Applications Electromobility Shift Assay, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 12H11
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG3 liquid in buffer containing 0.02MPhosphate buffer, 0.25MNaCl, pH 7.6 with 0.1% sodium azide.
Gene Accession No. NP_068810.3
Gene Symbols RELA, NFKB, p65, p50/p65
Host Species Mouse
Immunogen Peptide corresponding to human p65 coupled to BSA.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rabbit, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.