Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFĸB Mouse anti-Human, Mouse, Rat, Bovine, Clone: 1G10.2, MilliporeSigma™

Catalog No. 051469MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-146-9MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-146-9MI Supplier MilliporeSigma Supplier No. 051469
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-NFĸB Antibody, p65, clone 1G10.2 detects level of NFĸB and has been published and validated for use in western blot, immunofluorescence and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFκB
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1G10.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal in 0.1M Tris-Glycine (pH 7.4) with 150mM NaCl and 0.05% NaN3.
Gene Accession No. NP_068810.2
Host Species Mouse
Immunogen Synthetic peptide corresponding to the NLS region of human NFκB p65.
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.