Learn More
Description
Specifically detects NFκB p50 Clone: in Human samples, and it is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
| Antigen | NFκB p50 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. |
| Gene Accession No. | P19838 |
| Gene Symbols | NFKB1 |
| Host Species | Rabbit |
| Immunogen | KLH conjugated synthetic peptide (MAEDDPYLGRPE-C) corresponding to amino acids 1-12 of NFκB p50, containing a C-terminal cysteine. |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.