Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFκB p52 Mouse anti-Human, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05361
Description
Anti-NFκB p52 Antibody is a Mouse Monoclonal Antibody for detection of NFκB p52 also known as DNA-binding factor KBF2, Lymphocyte translocation chromosome 10, Oncogene Lyt-10 and has been validated in EMSA, immunoprecipitation and western blotting.
Specifications
NFκB p52 | |
Monoclonal | |
Purified mouse monoclonal IgG2a in buffer containing 0.1MTris-glycine, pH 7.4, 0.15MNaCl, 0.05% sodium azide. | |
NFKB2, LYT10, H2TF1, LYT-10, Lyt10 | |
GST-fusion protein corresponding to amino acids 1-444 of human NFκB p52 subunit (Schmidt, R., 1991;Bours, V., 1992). | |
200 μg | |
Human | |
Purified |
Electromobility Shift Assay, Immunoprecipitation, Western Blot | |
Unconjugated | |
NM_019408 | |
Mouse | |
Protein G chromatography | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction