Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NF-E2-related factor 2 Rabbit anti-Human, Mouse, Rat, Chimpanzee, Rhesus Macaque, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABE413
Description
Anti-NF-E2-related factor 2 Antibody is a highly specific rabbit polyclonal antibody, that targets NF-E2-related factor 2 and has been tested in western blotting and immunocytochemistry.
Specifications
NF-E2-related factor 2 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Chimpanzee, Human, Mouse, Rat, Rhesus Monkey |
Immunocytochemistry, Western Blot | |
Unconjugated | |
NFE2L2; NRF2 | |
KLH-conjugated linear peptide corresponding to the C-terminus of human NF-E2 related factor 2. | |
100 μg | |
NP_006155 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction