Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFAT-1 Mouse, Unlabeled, Clone: 1, BD

Catalog No. BDB610702 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610702 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610702 Supplier BD Biosciences Supplier No. 610702
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

T cells are activated and induced to proliferate following binding of their respective antigen. The process of includes expression of genes that encode factors (i.e., cytokines) which regulate various cell types. Modulation of gene expression is conducted by an array of specific interactions between transcription factors and DNA. NFAT-1 (Nuclear Factor of Activated T cells) is a transcription factor that regulates expression of the interleukin-2 gene. Thus, NFAT-1 DNA binding activity is undetectable in resting cells, but increases during T-cell activation. NFAT-1, a protein of 921 amino acids, is part of an oligomeric transcription factor that also contains Fra-1 and JunB. NFAT-1 was initially described as a phosphoprotein and is dephosphorylated in activated T cells transformed with the leukemia virus HTLV-l.

TRUSTED_SUSTAINABILITY

Specifications

Antigen NFAT-1
Applications Western Blot
Classification Monoclonal
Clone 1
Concentration 250μg/mL
Conjugate Unconjugated
Description NF-ATc2; NFATP; KIAA0611
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.