Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFAT1 Mouse anti-Human, Mouse, Rat, Clone: 8G4.2, MilliporeSigma™

Catalog No. 04804MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-804-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-804-MI Supplier MilliporeSigma Supplier No. 04804
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-NFAT1 Antibody, clone 8G4.2 is validated for use in western blot, immunofluorescence for the detection of NFAT1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFAT1
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 8G4.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal in PBS with 0.05% NaN3.
Gene Accession No. NP_001129493.1
Gene Symbols KIAA0611, NF-ATP , NF-ATp , NFAT1, NFATP, NFATp
Host Species Mouse
Immunogen GST-fusion protein corresponding to amino acids 1-297 of human NFAT1.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.