Learn More
Description
The product of this gene is a member of the nuclear factors of activated T cells family of transcription factors. Proteins belonging to this family play a central role in inducible gene transcription during the immune response. This protein regulates gene expression induced by osmotic stress in mammalian cells. Unlike monomeric members of this protein family, this protein exists as a homodimer and forms stable dimers with DNA elements. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
Specifications
| Antigen | NFAT5 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant NFAT5. |
| Formulation | 50% glycerol |
| Gene | NFAT5 |
| Gene Accession No. | NM_006599 |
| Gene Alias | KIAA0827/NF-AT5/NFATL1/NFATZ/OREBP/TONEBP |
| Gene Symbols | NFAT5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.