Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFATC2, Mouse, Clone: 2A4, Abnova™

Catalog No. 89123108 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-108 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-108 Supplier Abnova Corporation Supplier No. H00004773M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant NFATC2.

This gene is a member of the nuclear factor of activated T cells (NFAT) family. The product of this gene is a DNA-binding protein with a REL-homology region (RHR) and an NFAT-homology region (NHR). This protein is present in the cytosol and only translocates to the nucleus upon T cell receptor (TCR) stimulation, where it becomes a member of the nuclear factors of activated T cells transcription complex. This complex plays a central role in inducing gene transcription during the immune response. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq

Sequence: HYSPTNQQLRCGSHQEFQHIMYCENFAPGTTRPGPPPVSQGQRLSPGSYPTVIQQQNATSQRAAKNGPPVSDQKEVLPAGVTIKQEQNLDQTYLDDELIDTRLSWIQNIL

Specifications

Antigen NFATC2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2A4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant NFATC2.
Formulation ascites with no preservative
Gene NFATC2
Gene Accession No. NM_012340
Gene Alias KIAA0611/NFAT1/NFATP
Gene Symbols NFATC2
Host Species Mouse
Immunogen NFATC2 (NP_036472, 812 a.a. ∼ 921 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4773
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.