Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFATC2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89003043 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-003-043 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-003-043 Supplier Abnova Corporation Supplier No. H00004773A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant NFATC2.

This gene is a member of the nuclear factor of activated T cells (NFAT) family. The product of this gene is a DNA-binding protein with a REL-homology region (RHR) and an NFAT-homology region (NHR). This protein is present in the cytosol and only translocates to the nucleus upon T cell receptor (TCR) stimulation, where it becomes a member of the nuclear factors of activated T cells transcription complex. This complex plays a central role in inducing gene transcription during the immune response. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq

Sequence: HYSPTNQQLRCGSHQEFQHIMYCENFAPGTTRPGPPPVSQGQRLSPGSYPTVIQQQNATSQRAAKNGPPVSDQKEVLPAGVTIKQEQNLDQTYLDDELIDTRLSWIQNIL

Specifications

Antigen NFATC2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant NFATC2.
Formulation 50% glycerol
Gene NFATC2
Gene Accession No. NM_012340
Gene Alias KIAA0611/NFAT1/NFATP
Gene Symbols NFATC2
Host Species Mouse
Immunogen NFATC2 (NP_036472, 812 a.a. ∼ 921 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4773
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.