Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nuclear factor I/A, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017894 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-894 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-894 Supplier Abnova Corporation Supplier No. H00004774D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human NFIA protein.

Nuclear factor I (NFI) proteins constitute a family of dimeric DNA-binding proteins with similar, and possibly identical, DNA-binding specificity. They function as cellular transcription factors and as replication factors for adenovirus DNA replication. Diversity in this protein family is generated by multiple genes, differential splicing, and heterodimerization.[supplied by OMIM

Sequence: MYSPLCLTQDEFHPFIEALLPHVRAFAYTWFNLQARKRKYFKKHEKRMSKEEERAVKDELLSEKPEVKQKWASRLLAKLRKDIRPEYREDFVLTVTGKKPPCCVLSNPDQKGKMRRIDCLRQADKVWRLDLVMVILFKGIPLESTDGERLVKSPQCSNPGLCVQPHHIGVSVKELDLYLAYFVHAADSSQSESPSQPSDADIKDQPENGHLGFQDSFVTSGVFSVTELVRVSQTPIAAGTGPNFSLSDLESSSYYSMSPGAMRRSLPSTSSTSSTKRLKSVEDEMDSPGEEPFYTGQGRSPGSGSQSSGWHEVEPGMPSPTTLKKSEKSGFSSPSPSQTSSLGTAFTQHHRPVITGPRASPHATPSTLHFPTSPIIQQPGPYFSHPAIRYHPQETLKEFVQLVCPDAGQQAGQVGFLNPNGSSQGKVHNPFLPTPMLPPPPPPPMARPVPLPVPDTKPPTTSTEGGAASPTSPILVPGIKVAASHHPPDRPPDPFSTL

Specifications

Antigen nuclear factor I/A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human NFIA protein.
Formulation PBS with no preservative; pH 7.4
Gene NFIA
Gene Accession No. NM_005595.1
Gene Alias DKFZp434L0422/KIAA1439/NFI-L
Gene Symbols NFIA
Host Species Rabbit
Immunogen NFIA (NP_005586.1, 1 a.a. ∼ 498 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Gene ID (Entrez) 4774
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.