Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nuclear factor I/C (CCAAT-binding transcription factor), Mouse, Clone: 1D6, Abnova™

Catalog No. 89000834 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-834 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-834 Supplier Abnova Corporation Supplier No. H00004782M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant NFIC.

Sequence: TEDMEGGISSPVKKTEMDKSPFNSPSPQDSPRLSSFTQHHRPVIAVHSGIARSPHPSSALHFPTTSILPQTASTYFPHTAIRYPPHLNPQDPLKDLVSLACDPASQQPGP

Specifications

Antigen nuclear factor I/C (CCAAT-binding transcription factor)
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1D6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant NFIC.
Formulation PBS with no preservative; pH 7.4
Gene NFIC
Gene Accession No. NM_005597
Gene Alias CTF/CTF5/MGC20153/NF-I/NFI
Gene Symbols NFIC
Host Species Mouse
Immunogen NFIC (NP_005588, 314 a.a. ∼ 423 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 4782
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.