Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIK Rabbit anti-Human, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA515178
Description
NIK (mitogen-activated protein kinase kinase kinase 14), a member of the Ser/Thr protein kinase family, binds to TRAF2 and stimulates NF-kappaB activity. It shares sequence similarity with several other MAPKK kinases. It participates in an NF-kappaB-inducing signalling cascade common to receptors of the tumour-necrosis/nerve-growth factor (TNF/NGF) family and to the interleukin-1 type-I receptor.
Specifications
NIK | |
Polyclonal | |
MAP3K14 | |
Mitogen-activated protein kinase kinase kinase 14, NF-kappa beta-inducing kinase, Serine/threonine protein kinase NIK, HsNIK | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
360640, 9020 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q99558 | |
MAP3K14 | |
KLH conjugated synthetic peptide between 119-148 amino acids from the central region of human NIK | |
400 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction