Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFkB p65 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA535232
Description
This antibody is predicted to react with mouse based on sequence homology.
NFKB1 (MIM 164011) or NFKB2 (MIM 164012) is bound to REL(MIM 164910), RELA, or RELB (MIM 604758) to form the NFKB complex.The p50 (NFKB1)/p65 (RELA) heterodimer is the most abundant form ofNFKB. The NFKB complex is inhibited by I-kappa-B proteins (NFKBIA,MIM 164008 or NFKBIB, MIM 604495), which inactivate NFKB bytrapping it in the cytoplasm. Phosphorylation of serine residues onthe I-kappa-B proteins by kinases (IKBKA, MIM 600664, or IKBKB, MIM603258) marks them for destruction via the ubiquitination pathway,thereby allowing activation of the NFKB complex. Activated NFKBcomplex translocates into the nucleus and binds DNA atkappa-B-binding motifs such as 5-prime GGGRNNYYCC 3-prime or5-prime HGGARNYYCC 3-prime (where H is A, C, or T; R is an A or Gpurine; and Y is a C or T pyrimidine).
Specifications
| NFkB p65 | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| Q04206 | |
| RELA | |
| KLH conjugated synthetic peptide between 166-195 amino acids from the central region of human RELA | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| RELA | |
| RELA; NFKB3; Transcription factor p65; Nuclear factor NF-kappa-B p65 subunit; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5970 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction