Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFKBIL1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524176
Description
This gene encodes a divergent member of the I-kappa-B family of proteins. Its function has not been determined. The gene lies within the major histocompatibility complex (MHC) class I region on chromosome 6. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| NFKBIL1 | |
| Polyclonal | |
| NFKBIL1 | |
| IKBL, LST1, NFKBIL, I-kappa-B-like protein, NF-kappa-B inhibitor-like protein 1, ikappaBL, inhibitor of kappa B-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4795 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Q9UBC1 | |
| NFKBIL1 | |
| KLH conjugated synthetic peptide between 256-285 amino acids from the central region of human NFKBIL1 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction