Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nuclear transcription factor Y, gamma, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017898 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-898 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-898 Supplier Abnova Corporation Supplier No. H00004802D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human NFYC protein.

This gene encodes one subunit of a trimeric complex forming a highly conserved transcription factor that binds with high specificity to CCAAT motifs in the promoters of a variety of genes. The encoded protein, subunit C, forms a tight dimer with the B subunit, a prerequisite for subunit A association. The resulting trimer binds to DNA with high specificity and affinity. Subunits B and C each contain a histone-like motif. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MSTEGGFGGTSSSDAQQSLQSFWPRVMEEIRNLTVKDFRVQELPLARIKKIMKLDEDVKMISAEAPVLFAKAAQIFITELTLRAWIHTEDNKRRTLQRNDIAMAITKFDQFDFLIDIVPRDELKPPKRQEEVRQSVTPAEPVQYYFTLAQQPTAVQVQGQQQGQQTTSSTTTIQPGQIIIAQPQQGQTTPVTMQVGEGQQVQIVQAQPQGQAQQAQSGTGQTMQVMQQIITNTGEIQQIPVQLNAGQLQYIRLAQPVSGTQVVQGQIQTLATNAQQITQTEVQQGQQQFSQFTDGQQLYQIQQVTMPAGQDLAQPMFIQSANQPSDGQAPQVTGD

Specifications

Antigen nuclear transcription factor Y, gamma
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human NFYC protein.
Formulation PBS with no preservative; pH 7.4
Gene NFYC
Gene Accession No. NM_014223.2
Gene Alias CBF-C/CBFC/DKFZp667G242/FLJ45775/H1TF2A/HAP5/HSM/NF-YC
Gene Symbols NFYC
Host Species Rabbit
Immunogen NFYC (NP_055038.2, 1 a.a. ∼ 335 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Gene ID (Entrez) 4802
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.