Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NG2 Chondroitin Sulfate Proteoglycan Mouse anti-Human, Clone: 7.1, MilliporeSigma™

Catalog No. MAB5520MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5520MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5520MI Supplier MilliporeSigma Supplier No. MAB5520

Mouse Monoclonal Antibody

Anti-NG2 Chondroitin Sulfate Proteoglycan Antibody, clone 7.1 is an antibody against NG2 Chondroitin Sulfate Proteoglycan for use in FC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NG2 Chondroitin Sulfate Proteoglycan
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 7.1
Conjugate Unconjugated
Formulation Ascites fluid. Liquid. Contains no preservative.
Gene Accession No. NM_001897.4
Gene Symbols CSPG4, MSK16, MCSP, NG2, MCSPG, MEL-CSPG, HMW-MAA
Host Species Mouse
Immunogen Human bone marrow stromal cell line, SV40 transformed.
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.