Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NG2 Chondroitin Sulfate Proteoglycan, Cy3 Conjugate Rabbit anti-Rat, Mouse, Human, Monkey, Cy3, Polyclonal, MilliporeSigma™

Catalog No. AB5320C3MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5320C3MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5320C3MI Supplier MilliporeSigma Supplier No. AB5320C3
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-NG2 Chondroitin Sulfate Proteoglycan, Cy3 Conjugate Antibody is an antibody against NG2 Chondroitin Sulfate Proteoglycan for use in immunocytochemistry and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NG2 Chondroitin Sulfate Proteoglycan
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Conjugate Cyanine3
Formulation Purified rabbit polyclonal conjugated to Cy3 in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Symbols CSPG4; MCSP
Host Species Rabbit
Immunogen Immunoaffinity purified NG2 Chondroitin Sulfate Proteoglycan from rat
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_001888
Target Species Human, Monkey, Mouse, Rat
Content And Storage 2-8°C for 6 months in undiluted aliquotes.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.